missing translation for 'onlineSavingsMsg'
Learn More

Sodium perrhenate, 99+% (metals basis), Re 68%, Thermo Scientific Chemicals

Product Code. 11311229
Click to view available options
Quantity:
2 g
10 g
50 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Sodium perrhenate is used as a precursor to other rhenium compounds. Further, it reacts with sodium in ethanol to prepare nonahydridorhenate and with polysulfide solutions to get tetrathiorhenate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13472-33-8
Molecular Formula NaO4Re
Molecular Weight (g/mol) 273.193
MDL Number MFCD00014231
InChI Key KMISVFTVBOPTAI-UHFFFAOYSA-N
Synonym sodium perrhenate, unii-pe1t8nn47q, sodium tetraoxorhenate 1-, pe1t8nn47q, na.o4re, rhenium vii sodium oxide, rhenate reo41-, sodium, t-4, acmc-1bwm0, perrhenic acid sodium salt, sodium oxido trioxo rhenium
PubChem CID 5107658
IUPAC Name sodium;oxido(trioxo)rhenium
SMILES [O-][Re](=O)(=O)=O.[Na+]

Specifications

Melting Point 300°C
Color Yellow
Physical Form Crystalline
Quantity 50 g
Assay Percent Range 99+% (metals basis)
Solubility Information Soluble in water and ethanol.
Formula Weight 273.19
Odor Odorless
Sensitivity Hygroscopic
Appearance White crystalline powder
Density 5.39
Chemical Name or Material Sodium perrhenate, Re 68%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'