Learn More
Sodium perrhenate, 99+% (metals basis), Re 68%, Thermo Scientific Chemicals
£96.68 - £1727.60
Chemical Identifiers
CAS | 13472-33-8 |
---|---|
Molecular Formula | NaO4Re |
Molecular Weight (g/mol) | 273.193 |
MDL Number | MFCD00014231 |
InChI Key | KMISVFTVBOPTAI-UHFFFAOYSA-N |
Synonym | sodium perrhenate, unii-pe1t8nn47q, sodium tetraoxorhenate 1-, pe1t8nn47q, na.o4re, rhenium vii sodium oxide, rhenate reo41-, sodium, t-4, acmc-1bwm0, perrhenic acid sodium salt, sodium oxido trioxo rhenium |
PubChem CID | 5107658 |
IUPAC Name | sodium;oxido(trioxo)rhenium |
SMILES | [O-][Re](=O)(=O)=O.[Na+] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11331229
|
Thermo Scientific Alfa Aesar
011412.04 |
2 g |
£96.68
2g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11321229
|
Thermo Scientific Alfa Aesar
011412.09 |
10 g |
£402.00
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11311229
|
Thermo Scientific Alfa Aesar
011412.18 |
50 g |
£1727.60
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Sodium perrhenate is used as a precursor to other rhenium compounds. Further, it reacts with sodium in ethanol to prepare nonahydridorhenate and with polysulfide solutions to get tetrathiorhenate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
13472-33-8 | |
273.193 | |
KMISVFTVBOPTAI-UHFFFAOYSA-N | |
5107658 | |
[O-][Re](=O)(=O)=O.[Na+] |
NaO4Re | |
MFCD00014231 | |
sodium perrhenate, unii-pe1t8nn47q, sodium tetraoxorhenate 1-, pe1t8nn47q, na.o4re, rhenium vii sodium oxide, rhenate reo41-, sodium, t-4, acmc-1bwm0, perrhenic acid sodium salt, sodium oxido trioxo rhenium | |
sodium;oxido(trioxo)rhenium |
Specifications
300°C | |
Yellow | |
99+% (metals basis) | |
MFCD00014231 | |
Soluble in water and ethanol. | |
[O-][Re](=O)(=O)=O.[Na+] | |
273.193 | |
273.19 | |
Hygroscopic | |
5.39 |
13472-33-8 | |
Crystalline | |
NaO4Re | |
sodium perrhenate, unii-pe1t8nn47q, sodium tetraoxorhenate 1-, pe1t8nn47q, na.o4re, rhenium vii sodium oxide, rhenate reo41-, sodium, t-4, acmc-1bwm0, perrhenic acid sodium salt, sodium oxido trioxo rhenium | |
KMISVFTVBOPTAI-UHFFFAOYSA-N | |
sodium;oxido(trioxo)rhenium | |
5107658 | |
Odorless | |
White crystalline powder | |
Sodium perrhenate, Re 68% |
Safety and Handling
P210-P220-P221-P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362-P370+P378q-P501c
H272-H315-H319
missing translation for 'dotInformation' : DOT Class: 5.1, Packaging Group: II
missing translation for 'einecsNumber' : 236-742-9
missing translation for 'rtecsNumber' : WD3675000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Sodium perrhenate
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 2
- Oxidising solid Category 2
- Skin corrosion/irritation Category 2
Hazard Statement
- H272-May intensify fire; oxidiser.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P371+P380+P375-In case of major fire and large quantities: Evacuate area Fight fire remotely due to the risk of explosion.
Supplemental information
- MIXTURE LIST-Contains : Sodium perrhenate
RUO – Research Use Only