missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Persulfate, Honeywell
Click to view available options
Quantity:
500 g
1 kg
2.5 kg
5 kg
Description
- ≥95% for general laboratory use.
Specifications
Specifications
Quantity | 1 kg |
Linear Formula | Na2S2O8 |
UN Number | 1505 |
Formula Weight | 238.1g/mol |
Percent Purity | ≥98% |
Grade | Reagent |
Chemical Name or Material | Sodium Persulfate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'