Learn More
Thermo Scientific Chemicals Sodium phosphate, 0.5M buffer soln., pH 7.0
Brand: Thermo Scientific Chemicals J63791.AP
97.20 GBP valid until 2024-12-20
Use promo code "21651" to get your promotional price.
Description
Sodium phosphate is widely used in molecular biology, biochemistry, and chromatography. Useful in conjunction with Sodium Phosphate, Monobasic in the preparation of biological buffers. Used in many applications including the purification of antibodies.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsSodium phosphate is widely used in molecular biology, biochemistry, and chromatography. Useful in conjunction with Sodium Phosphate, Monobasic in the preparation of biological buffers. Used in many applications including the purification of antibodies.
Solubility
Fully miscible.
Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
Specifications
Sodium phosphate buffer soln. | |
7.0 | |
500 mL | |
Fully miscible. |
0.5 M | |
Liquid | |
MFCD00146206 |