missing translation for 'onlineSavingsMsg'
Learn More

Sodium Phosphate, Dibasic, 7-Hydrate, Endotoxin Tested, Crystal, U.S.P., J.T. Baker™

Product Code. 15548174
Click to view available options
Quantity:
2.5 kg
Packaging:
Poly Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Specifications

CAS 7782-85-6
Physical Form Crystals
Quantity 2.5 kg
Molecular Formula H17Na2O11P
MDL Number MFCD00149180
Synonym sodium phosphate dibasic heptahydrate, unii-70wt22sf4b, sodium phosphate, dibasic, heptahydrate, sodium hydrogen phosphate heptahydrate, disodium hydrogen phosphate heptahydrate, axea paragraph signthornaae, 2na.hpo4.7h2o, sodium hydrogenphosphate heptahydrate, sodium phosphate dibasic, acs, disodium hydrogenphosphate heptahydrate
InChI Key PYLIXCKOHOHGKQ-UHFFFAOYSA-N
SMILES O.O.O.O.O.O.O.[Na+].[Na+].OP(O)(O)=O
IUPAC Name disodium phosphoric acid heptahydrate
Molecular Weight (g/mol) 270.08
PubChem CID 6096963
Formula Weight 268.07
Grade USP
Packaging Poly Bottle
Specific Gravity 1L = 1.70kg
Chemical Name or Material Sodium Phosphate, Dibasic, 7-Hydrate, Endotoxin Tested
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'