missing translation for 'onlineSavingsMsg'
Learn More

Sodium phosphate dibasic, ACS Reagent, Honeywell Fluka™

Catalog Number 15698160
Click to view available options
Quantity:
100 g
500 g
1 kg
2.5 kg
5 kg
6 x 500 g
12 kg
4 x 2.5 kg
This item is not returnable. View return policy
7558-79-4
HNa2O4P
141.96
MFCD00003496
BNIILDVGGAEEIG-UHFFFAOYSA-L
disodium hydrogen phosphate, disodium phosphate, disodium hydrogenorthophosphate, sodium phosphate dibasic, dibasic sodium phosphate, acetest, soda phosphate, disodium orthophosphate, sodium hydrogen phosphate, phosphoric acid, disodium salt
24203
CHEBI:34683
[Na+].[Na+].OP([O-])([O-])=O
This item is not returnable. View return policy
7558-79-4
HNa2O4P
141.96
MFCD00003496
BNIILDVGGAEEIG-UHFFFAOYSA-L
disodium hydrogen phosphate, disodium phosphate, disodium hydrogenorthophosphate, sodium phosphate dibasic, dibasic sodium phosphate, acetest, soda phosphate, disodium orthophosphate, sodium hydrogen phosphate, phosphoric acid, disodium salt
24203
CHEBI:34683
[Na+].[Na+].OP([O-])([O-])=O

ACS Reagent,≥99.0%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY
Quantity 5 kg
Linear Formula Na2HPO4
UN Number NONH for all modes of transport
IUPAC Name disodium;hydrogen phosphate
Formula Weight 141.96g/mol
Percent Purity ≥99%
Grade ACS Reagent
Chemical Name or Material Sodium phosphate dibasic
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'