missing translation for 'onlineSavingsMsg'
Learn More

Sodium Phosphate Dibasic Anhydrous (White Granules or Powder), Fisher BioReagents™

Product Code. 10182863
Click to view available options
Quantity:
500 g
1 kg
Packaging:
Glass Bottle
Poly Bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21626” to get your promotional price
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21626” to get your promotional price

Sodium Phosphate Dibasic Anhydrous (White Granules or Powder)

This buffer component is commonly used in tissue culture and molecular biology applications.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7558-79-4
Molecular Formula HNa2O4P
Molecular Weight (g/mol) 141.96
MDL Number MFCD00003496
InChI Key BNIILDVGGAEEIG-UHFFFAOYSA-L
Synonym disodium hydrogen phosphate, disodium phosphate, disodium hydrogenorthophosphate, sodium phosphate dibasic, dibasic sodium phosphate, acetest, soda phosphate, disodium orthophosphate, sodium hydrogen phosphate, phosphoric acid, disodium salt
PubChem CID 24203
ChEBI CHEBI:34683
SMILES [Na+].[Na+].OP([O-])([O-])=O

Specifications

Color White
pH 8.7 to 9.3
Physical Form Powder/Solid
Quantity 500 g
Assay Percent Range ≥99.0 %
Merck Index 15, 8789
IUPAC Name disodium;hydrogen phosphate
Formula Weight 141.96
Percent Purity ≥99.0%
Identification Pass Test
Loss on Drying 0.2% max.
Packaging Glass Bottle
Decomposition Information Decomposes information: > 240°C
Chemical Name or Material Sodium Phosphate Dibasic Anhydrous
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'