missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium phosphate dibasic dihydrate, 98+%, for analysis, Thermo Scientific Chemicals
£21.91 - £51.56
Chemical Identifiers
CAS | 10028-24-7 |
---|---|
Molecular Formula | H5Na2O6P |
Molecular Weight (g/mol) | 177.99 |
MDL Number | MFCD00149182 |
InChI Key | KDQPSPMLNJTZAL-UHFFFAOYSA-L |
Synonym | disodium hydrogen phosphate dihydrate, disodium phosphate dihydrate, sodium phosphate dibasic dihydrate, di-sodium hydrogen phosphate dihydrate, phosphoric acid, disodium salt, dihydrate, sodium hydrogenphosphate dihydrate, sodium phosphate, dibasic, dihydrate, disodium hydrogenphosphate dihydrate, dinatrii phosphas dihydricus, 2na.hpo4.2h2o |
PubChem CID | 6451167 |
IUPAC Name | disodium;hydrogen phosphate;dihydrate |
SMILES | O.O.[Na+].[Na+].OP([O-])([O-])=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
12644977
|
Thermo Scientific Acros
124992500 |
250 g | Glass bottle |
£21.91
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12694947
|
Thermo Scientific Acros
124990010 |
1 kg | Glass bottle |
£51.56
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
10028-24-7 | |
177.99 | |
KDQPSPMLNJTZAL-UHFFFAOYSA-L | |
6451167 | |
O.O.[Na+].[Na+].OP([O-])([O-])=O |
H5Na2O6P | |
MFCD00149182 | |
disodium hydrogen phosphate dihydrate, disodium phosphate dihydrate, sodium phosphate dibasic dihydrate, di-sodium hydrogen phosphate dihydrate, phosphoric acid, disodium salt, dihydrate, sodium hydrogenphosphate dihydrate, sodium phosphate, dibasic, dihydrate, disodium hydrogenphosphate dihydrate, dinatrii phosphas dihydricus, 2na.hpo4.2h2o | |
disodium;hydrogen phosphate;dihydrate |
Specifications
243.0°C to 245.0°C | |
98+% | |
Na2HPO4·2H2O | |
15,8789 | |
(5% in water) Clear colorless | |
O.O.[Na+].[Na+].OP([O-])([O-])=O | |
177.99 | |
177.99 | |
Analytical |
10028-24-7 | |
H5Na2O6P | |
MFCD00149182 | |
disodium hydrogen phosphate dihydrate, disodium phosphate dihydrate, sodium phosphate dibasic dihydrate, di-sodium hydrogen phosphate dihydrate, phosphoric acid, disodium salt, dihydrate, sodium hydrogenphosphate dihydrate, sodium phosphate, dibasic, dihydrate, disodium hydrogenphosphate dihydrate, dinatrii phosphas dihydricus, 2na.hpo4.2h2o | |
KDQPSPMLNJTZAL-UHFFFAOYSA-L | |
disodium;hydrogen phosphate;dihydrate | |
6451167 | |
≥98% | |
Sodium phosphate dibasic dihydrate |
RUO – Research Use Only