missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium phosphate dibasic dihydrate, meets analytical spec. of BP, Ph. Eur., 98.5-101% (calc. to the dried substance), Honeywell Fluka™
Click to view available options
Quantity:
1 kg
2.5 kg
5 kg
25 kg
4 x 5 kg
6 x 1 kg
6 x 2.5 kg
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
CAS Max % | 100.0000% |
Quantity | 25 kg |
Assay Percent Range | 98.5 to 101% |
Linear Formula | Na2HPO4 · 2H2O |
IUPAC Name | disodium;hydrogen phosphate;dihydrate |
Formula Weight | 177.99 |
Percent Purity | 98.5 to 101% (calc. to the dried substance) |
Grade | Reagent Ph. Eur. |
Loss on Drying | 19.5 - 21.0% |
Packaging | Paper / Poly Bag |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'