missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Sodium phosphate, dibasic dodecahydrate, 99%, for biochemistry

Product Code. 13585000 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
2.5 kg
Unit Size:
2.5kg
This item is not returnable. View return policy

Product Code. 13585000

Brand: Thermo Scientific Chemicals 447250025

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10039-32-4 | HNa2O412H2O | 358.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Melting Point 35.0°C
CAS 10039-32-4
Quantity 2.5 kg
Assay Percent Range 99%
Molecular Formula HNa2O412H2O
Linear Formula Na2HPO4·12H2O
Merck Index 15,8789
Synonym disodium phosphate dodecahydrate, disodium hydrogen phosphate dodecahydrate, phosphoric acid, disodium salt, dodecahydrate, di-sodium hydrogen phosphate dodecahydrate, sodium hydrogenphosphate dodecahydrate, unii-e1w4n241fo, sodium phosphate dibasic dodecahydrate, sodium phosphate, dibasic, dodecahydrate, disodium orthophosphate, dodecahydrate, sodium monohydrogen phosphate dodecahydrate 2:1:1:12
Solubility Information Clear colorless
InChI Key DGLRDKLJZLEJCY-UHFFFAOYSA-L
SMILES O.O.O.O.O.O.O.O.O.O.O.O.OP(=O)([O-])[O-].[Na+].[Na+]
IUPAC Name disodium;hydrogen phosphate;dodecahydrate
Molecular Weight (g/mol) 358.13
PubChem CID 61456
Formula Weight 358.13
Percent Purity 99%
Grade Biochemical
Packaging Plastic bottle
Chemical Name or Material Sodium phosphate, dibasic dodecahydrate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.