missing translation for 'onlineSavingsMsg'
Learn More

Sodium phosphate, dibasic dodecahydrate, 99%, extra pure, Thermo Scientific Chemicals

£44.62 - £162.81

Chemical Identifiers

CAS 10039-32-4
Molecular Formula HNa2O412H2O
Molecular Weight (g/mol) 358.13
InChI Key DGLRDKLJZLEJCY-UHFFFAOYSA-L
Synonym disodium phosphate dodecahydrate, disodium hydrogen phosphate dodecahydrate, phosphoric acid, disodium salt, dodecahydrate, di-sodium hydrogen phosphate dodecahydrate, sodium hydrogenphosphate dodecahydrate, unii-e1w4n241fo, sodium phosphate dibasic dodecahydrate, sodium phosphate, dibasic, dodecahydrate, disodium orthophosphate, dodecahydrate, sodium monohydrogen phosphate dodecahydrate 2:1:1:12
PubChem CID 61456
IUPAC Name disodium;hydrogen phosphate;dodecahydrate
SMILES O.O.O.O.O.O.O.O.O.O.O.O.OP(=O)([O-])[O-].[Na+].[Na+]
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
12695147
View Documents Promotion Details
Thermo Scientific Acros
448160010
1 kg Plastic bottle
£44.62
1kg
Please to purchase this item. Need a web account? Register with us today!
12605157
View Documents
Thermo Scientific Acros
448160050
5 kg Plastic bottle
£162.81
5kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

10039-32-4
358.13
disodium phosphate dodecahydrate, disodium hydrogen phosphate dodecahydrate, phosphoric acid, disodium salt, dodecahydrate, di-sodium hydrogen phosphate dodecahydrate, sodium hydrogenphosphate dodecahydrate, unii-e1w4n241fo, sodium phosphate dibasic dodecahydrate, sodium phosphate, dibasic, dodecahydrate, disodium orthophosphate, dodecahydrate, sodium monohydrogen phosphate dodecahydrate 2:1:1:12
disodium;hydrogen phosphate;dodecahydrate
HNa2O412H2O
DGLRDKLJZLEJCY-UHFFFAOYSA-L
61456
O.O.O.O.O.O.O.O.O.O.O.O.OP(=O)([O-])[O-].[Na+].[Na+]

Specifications

35.0°C
99%
Na2HPO4·12H2O
disodium phosphate dodecahydrate, disodium hydrogen phosphate dodecahydrate, phosphoric acid, disodium salt, dodecahydrate, di-sodium hydrogen phosphate dodecahydrate, sodium hydrogenphosphate dodecahydrate, unii-e1w4n241fo, sodium phosphate dibasic dodecahydrate, sodium phosphate, dibasic, dodecahydrate, disodium orthophosphate, dodecahydrate, sodium monohydrogen phosphate dodecahydrate 2:1:1:12
DGLRDKLJZLEJCY-UHFFFAOYSA-L
disodium;hydrogen phosphate;dodecahydrate
61456
99%
Sodium phosphate, dibasic dodecahydrate
10039-32-4
HNa2O412H2O
15,8789
Solubility in water: 218g/L (20°C). Other solubilities: practically insoluble in alcohol
O.O.O.O.O.O.O.O.O.O.O.O.OP(=O)([O-])[O-].[Na+].[Na+]
358.13
358.13
Extra Pure
Videos
SDS
Documents

Documents

Product Certifications

RUO – Research Use Only