missing translation for 'onlineSavingsMsg'
Learn More

Sodium phosphate dibasic dodecahydrate, ≥99% (T), Honeywell™

Product Code. 15636810 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
1 kg
Unit Size:
1kg
This item is not returnable. View return policy

Product Code. 15636810

Brand: Honeywell-Fluka 716501KG

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Puriss. p.a., crystallized,≥99.0% (T)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Specifications

CAS 10039-32-4
Quantity 1 kg
Molecular Formula H25Na2O16P
Linear Formula Na2HPO4 · 12H2O
MDL Number MFCD00149181
Synonym disodium phosphate dodecahydrate, disodium hydrogen phosphate dodecahydrate, phosphoric acid, disodium salt, dodecahydrate, di-sodium hydrogen phosphate dodecahydrate, sodium hydrogenphosphate dodecahydrate, unii-e1w4n241fo, sodium phosphate dibasic dodecahydrate, sodium phosphate, dibasic, dodecahydrate, disodium orthophosphate, dodecahydrate, sodium monohydrogen phosphate dodecahydrate 2:1:1:12
InChI Key DGLRDKLJZLEJCY-UHFFFAOYSA-L
SMILES O.O.O.O.O.O.O.O.O.O.O.O.OP(=O)([O-])[O-].[Na+].[Na+]
IUPAC Name disodium;hydrogen phosphate;dodecahydrate
Molecular Weight (g/mol) 358.137
PubChem CID 61456
Formula Weight 358.14g/mol
Percent Purity ≥99% (T)
Chemical Name or Material Sodium phosphate dibasic dodecahydrate
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.