missing translation for 'onlineSavingsMsg'
Learn More

Sodium phosphate dibasic dodecahydrate, Puriss. p.a., crystallized, ≥99.0% (T), Honeywell Fluka™

Catalog Number 16665913
Click to view available options
10 kg
This item is not returnable. View return policy
This item is not returnable. View return policy
Melting Point 34°C
CAS 10039-32-4
Color Colorless
CAS Max % 1
pH 9 to 9.3
Quantity 10 kg
Molecular Formula H25Na2O16P
Linear Formula Na2HPO4 · 12H2O
Synonym sec-Sodium phosphate; Disodium hydrogen phosphate dodecahydrate; Disodium phosphate; di-Sodium hydrogen phosphate dodecahydrate
InChI Key DGLRDKLJZLEJCY-UHFFFAOYSA-L
SMILES O.O.O.O.O.O.O.O.O.O.O.O.[Na+].[Na+].OP([O-])([O-])=O
IUPAC Name disodium dodecahydrate hydrogen phosphate
Molecular Weight (g/mol) 358.14
Grade ACS
Density 1.52 g/cm3 (20°C)
Chemical Name or Material Sodium phosphate dibasic dodecahydrate
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'