missing translation for 'onlineSavingsMsg'
Learn More
Sodium phosphate, dibasic heptahydrate, 99+%, for analysis, Thermo Scientific Chemicals Product Code.: 11904291

Sodium phosphate, dibasic heptahydrate, 99+%, for analysis, Thermo Scientific Chemicals

Product Code. 11904291
2.5 kg, Plastic bottle
Click to view available options
Quantity:
250 g
500 g
2.5 kg
Packaging:
Plastic bottle
This item is not returnable. View return policy

Product Code. 11904291

Brand: Thermo Scientific Acros 206510025

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Biological buffer
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7782-85-6
Molecular Formula H15Na2O11P
Molecular Weight (g/mol) 268.06
MDL Number MFCD00149180
InChI Key LVXHNCUCBXIIPE-UHFFFAOYSA-L
Synonym sodium phosphate dibasic heptahydrate, unii-70wt22sf4b, sodium phosphate, dibasic, heptahydrate, sodium hydrogen phosphate heptahydrate, disodium hydrogen phosphate heptahydrate, axea paragraph signthornaae, 2na.hpo4.7h2o, sodium hydrogenphosphate heptahydrate, sodium phosphate dibasic, acs, disodium hydrogenphosphate heptahydrate
PubChem CID 6096963
IUPAC Name disodium phosphoric acid heptahydrate
SMILES O.[Na+].[Na+].OP([O-])([O-])=O

Specifications

Melting Point 48.0°C
Color White
pH 9.0 to 9.3 (5% soln. at 20°C)
Physical Form Crystals
Quantity 2.5 kg
Assay Percent Range 99+%
Linear Formula Na2HPO4·7H2O
Merck Index 15,8789
Solubility Information Solubility in water: 154g/L (20°C). Other solubilities: practically insoluble in alcohol
Formula Weight 268.07
Percent Purity ≥99%
Grade Analytical
Packaging Plastic bottle
Density 1.67
Chemical Name or Material Sodium phosphate, dibasic heptahydrate, p.a.
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sodium phosphate, dibasic heptahydrate, 99+%, for analysis, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.