missing translation for 'onlineSavingsMsg'
Learn More

Sodium phosphate dibasic, puriss p.a. ACS Reagent, Honeywell Fluka™

Catalog Number 15616810
Click to view available options
Quantity:
250 g
1 kg
5 kg
7558-79-4
HNa2O4P
141.96
MFCD00003496
BNIILDVGGAEEIG-UHFFFAOYSA-L
disodium hydrogen phosphate, disodium phosphate, disodium hydrogenorthophosphate, sodium phosphate dibasic, dibasic sodium phosphate, acetest, soda phosphate, disodium orthophosphate, sodium hydrogen phosphate, phosphoric acid, disodium salt
24203
CHEBI:34683
disodium;hydrogen phosphate
[Na+].[Na+].OP([O-])([O-])=O
7558-79-4
HNa2O4P
141.96
MFCD00003496
BNIILDVGGAEEIG-UHFFFAOYSA-L
disodium hydrogen phosphate, disodium phosphate, disodium hydrogenorthophosphate, sodium phosphate dibasic, dibasic sodium phosphate, acetest, soda phosphate, disodium orthophosphate, sodium hydrogen phosphate, phosphoric acid, disodium salt
24203
CHEBI:34683
disodium;hydrogen phosphate
[Na+].[Na+].OP([O-])([O-])=O

Puriss. p.a., ACS Reagent, anhydrous,≥99.0% (T)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY
Quantity 250 g
Linear Formula Na2HPO4
Formula Weight 141.96g/mol
Percent Purity ≥99% (T)
Grade ACS/puriss. p.a.
Chemical Name or Material Sodium phosphate dibasic
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'