missing translation for 'onlineSavingsMsg'
Learn More
Sodium phosphate, monobasic dihydrate, 98%, extra pure, Thermo Scientific Chemicals Product Code.: 12645107

Sodium phosphate, monobasic dihydrate, 98%, extra pure, Thermo Scientific Chemicals

Product Code. 12645107
1 kg
Click to view available options
Quantity:
1 kg
5 kg
25 kg
This item is not returnable. View return policy

Product Code. 12645107

Brand: Thermo Scientific Acros 447760010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13472-35-0
Molecular Formula H2NaO42H2O
Molecular Weight (g/mol) 156.01
InChI Key VBJGJHBYWREJQD-UHFFFAOYSA-M
Synonym sodium dihydrogen phosphate dihydrate, sodium phosphate monobasic dihydrate, monosodium phosphate dihydrate, sodium phosphate, monobasic, dihydrate, phosphoric acid, monosodium salt, dihydrate, sodium dihydrogenphosphate dihydrate, sodium phosphate nah2po4 dihydrate, monosodium dihydrogen phosphate dihydrate, sodium dihydrate dihydrogen phosphate, acmc-20ajoh
PubChem CID 23673460
IUPAC Name sodium;dihydrogen phosphate;dihydrate
SMILES O.O.OP(=O)(O)[O-].[Na+]

Specifications

Melting Point 60.0°C
Quantity 1 kg
Assay Percent Range 98%
Linear Formula NaH2PO4·2H2O
Merck Index 15,879
Solubility Information Solubility in water: soluble. Other solubilities: practically insoluble in alcohol
Formula Weight 156.01
Percent Purity 98%
Grade Extra Pure
Packaging Plastic bottle
Chemical Name or Material Sodium phosphate, monobasic dihydrate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sodium phosphate, monobasic dihydrate, 98%, extra pure, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.