missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium phosphate, monobasic monohydrate, 98%, extra pure, Thermo Scientific Chemicals
missing translation for 'orderingAttributeHoverText'
Quantity:
1 kg
5 kg
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Spécification
Melting Point | 100.0°C |
Quantity | 1 kg |
Assay Percent Range | 98% |
Linear Formula | NaH2PO4·H2O |
Merck Index | 15,879 |
Solubility Information | Solubility in water: 1103g/L (20°C). Other solubilities: practically insoluble in alcohol |
IUPAC Name | sodium;dihydrogen phosphate;hydrate |
Formula Weight | 137.99 |
Percent Purity | 98% |
Grade | Extra Pure |
Afficher plus |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'