missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Phosphate Monobasic Monohydrate (Colorless-to-white Crystals), Fisher BioReagents™
Click to view available options
Quantity:
500 g
1 kg
Packaging:
Amber Glass
Description
Sodium Phosphate Monobasic Monohydrate (Colorless-to-white Crystals)
Sodium Phosphate is commonly used in tissue culture and molecular biology applications.
Specifications
Melting Point | 100°C |
Color | White |
pH | 4.1 to 4.5 |
Physical Form | Solid |
Quantity | 500 g |
IUPAC Name | sodium;dihydrogen phosphate;hydrate |
Formula Weight | 137.99 |
Percent Purity | 98.0 to 102.0% |
Identification | Pass Test |
Packaging | Amber Glass |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'