missing translation for 'onlineSavingsMsg'
Learn More
Sodium Phosphate, Monobasic, Monohydrate, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™ Product Code.: 15382268
compliance-icons

Sodium Phosphate, Monobasic, Monohydrate, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™

Product Code. 15382268
500 g, Poly Bottle
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
This item is not returnable. View return policy

Product Code. 15382268

Brand: Avantor J.T.Baker 381801

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High quality chemicals for laboratory and specialized industrial use

Chemical Identifiers

CAS 10049-21-5
Molecular Formula H5NaO5P
Molecular Weight (g/mol) 139.00
MDL Number MFCD00149208
InChI Key BBMHARZCALWXSL-UHFFFAOYSA-N
Synonym sodium dihydrogen phosphate monohydrate, sodium phosphate monobasic monohydrate, monosodium phosphate monohydrate, unii-593yog76rn, phosphoric acid, monosodium salt, monohydrate, sodium phosphate monobasic hydrate, sodium dihydrogen phosphate hydrate, sodium dihydrogenphosphate monohydrate, sodium hydrate dihydrogen phosphate, pubchem12707
PubChem CID 516949
SMILES O.[Na+].OP(O)(O)=O

Specifications

Physical Form Crystals
Quantity 500 g
IUPAC Name sodium;dihydrogen phosphate;hydrate
Formula Weight 137.99
Grade BAKER ANALYZED™ ACS Reagent
Packaging Poly Bottle
Specific Gravity 1L = 2.04kg
Chemical Name or Material Sodium Phosphate, Monobasic, Monohydrate
compliance-icons
Product Identifier
  • Sodium Phosphate, Monobasic, Monohydrate, Crystal
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P264-Wash hands thoroughly after handling.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : sodium phosphate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sodium Phosphate, Monobasic, Monohydrate, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.