missing translation for 'onlineSavingsMsg'
Learn More
Sodium phosphate, tribasic dodecahydrate, 98+%, ACS reagent, Thermo Scientific Chemicals Product Code.: 10582364

Sodium phosphate, tribasic dodecahydrate, 98+%, ACS reagent, Thermo Scientific Chemicals

Product Code. 10582364
2.5 kg, Plastic bottle
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Plastic bottle
This item is not returnable. View return policy

Product Code. 10582364

Brand: Thermo Scientific Acros 424400025

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10101-89-0
Molecular Formula Na3O412H2O
Molecular Weight (g/mol) 380.13
MDL Number MFCD00149198
InChI Key ASTWEMOBIXQPPV-UHFFFAOYSA-K
Synonym trisodium phosphate dodecahydrate, sodium phosphate dodecahydrate, sodium phosphate tribasic dodecahydrate, phosphoric acid, trisodium salt, dodecahydrate, unii-b70850qphr, ccris 7322, sodium phosphate, tribasic, dodecahydrate, trisodium dodecahydrate phosphate, phosphoric acid, trisodium salt, dodeahydrate, trisodium phosphate tert dodecahydrate
PubChem CID 61473
IUPAC Name trisodium;phosphate;dodecahydrate
SMILES O.O.O.O.O.O.O.O.O.O.O.O.[O-]P(=O)([O-])[O-].[Na+].[Na+].[Na+]

Specifications

Melting Point 75.0°C
Color White
Physical Form Crystalline Powder or Needles
Quantity 2.5 kg
Assay Percent Range 98+%
Linear Formula Na3PO4·12H2O
Merck Index 15, 8792
Solubility Information Solubility in water: 240g/L (20°C). Other solubilities: practically insoluble in alcohol
Formula Weight 380.13
Percent Purity ≥98%
Grade ACS Reagent
Packaging Plastic bottle
Specific Gravity 1.62
Density 1.6200g/mL
Chemical Name or Material Sodium phosphate, tribasic dodecahydrate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sodium phosphate, tribasic dodecahydrate, 98+%, ACS reagent, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.