Learn More
Sodium phosphate, tribasic dodecahydrate, 98%, for analysis, Thermo Scientific Chemicals
£67.54 - £728.63
Chemical Identifiers
CAS | 10101-89-0 |
---|---|
Molecular Formula | Na3O4P·12H2O |
Molecular Weight (g/mol) | 380.13 |
MDL Number | MFCD00149198 |
InChI Key | ASTWEMOBIXQPPV-UHFFFAOYSA-K |
Synonym | trisodium phosphate dodecahydrate, sodium phosphate dodecahydrate, sodium phosphate tribasic dodecahydrate, phosphoric acid, trisodium salt, dodecahydrate, unii-b70850qphr, ccris 7322, sodium phosphate, tribasic, dodecahydrate, trisodium dodecahydrate phosphate, phosphoric acid, trisodium salt, dodeahydrate, trisodium phosphate tert dodecahydrate |
PubChem CID | 61473 |
IUPAC Name | trisodium;phosphate;dodecahydrate |
SMILES | O.O.O.O.O.O.O.O.O.O.O.O.[O-]P(=O)([O-])[O-].[Na+].[Na+].[Na+] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10082750
|
Thermo Scientific Acros
206520010 |
1 kg | Plastic bottle |
£67.54
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10115693
|
Thermo Scientific Acros
206520025 |
2.5 kg | Plastic bottle |
£140.70
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10667934
|
Thermo Scientific Acros
206520251 |
25 kg | Fibre drum |
£728.63
25kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Biological bufferChemical Identifiers
10101-89-0 | |
380.13 | |
ASTWEMOBIXQPPV-UHFFFAOYSA-K | |
61473 | |
O.O.O.O.O.O.O.O.O.O.O.O.[O-]P(=O)([O-])[O-].[Na+].[Na+].[Na+] |
Na3O4P·12H2O | |
MFCD00149198 | |
trisodium phosphate dodecahydrate, sodium phosphate dodecahydrate, sodium phosphate tribasic dodecahydrate, phosphoric acid, trisodium salt, dodecahydrate, unii-b70850qphr, ccris 7322, sodium phosphate, tribasic, dodecahydrate, trisodium dodecahydrate phosphate, phosphoric acid, trisodium salt, dodeahydrate, trisodium phosphate tert dodecahydrate | |
trisodium;phosphate;dodecahydrate |
Specifications
75.0°C | |
White | |
98% | |
Na3PO4·12H2O | |
15, 8792 | |
Solubility in water: 240g/L (20°C). Other solubilities: practically insoluble in alcohol | |
O.O.O.O.O.O.O.O.O.O.O.O.[O-]P(=O)([O-])[O-].[Na+].[Na+].[Na+] | |
380.13 | |
380.13 | |
Analytical | |
1.6200g/mL |
10101-89-0 | |
Crystalline Powder, Needles or Crystals | |
Na3O4P·12H2O | |
MFCD00149198 | |
trisodium phosphate dodecahydrate, sodium phosphate dodecahydrate, sodium phosphate tribasic dodecahydrate, phosphoric acid, trisodium salt, dodecahydrate, unii-b70850qphr, ccris 7322, sodium phosphate, tribasic, dodecahydrate, trisodium dodecahydrate phosphate, phosphoric acid, trisodium salt, dodeahydrate, trisodium phosphate tert dodecahydrate | |
ASTWEMOBIXQPPV-UHFFFAOYSA-K | |
trisodium;phosphate;dodecahydrate | |
61473 | |
98% | |
1.62 | |
Sodium phosphate, tribasic dodecahydrate |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Avoid breath
GHS Signal Word: Warning
- Sodium phosphate, tribasic dodecahydrate
Signal Word
- Warning
Hazard Category
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
- MIXTURE LIST-Contains : Tribasic sodium phosphate dodecahydrate,Trisodium phosphate
RUO – Research Use Only