missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium phosphite dibasic pentahydrate, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Quantity | 1 kg |
Assay Percent Range | 98% |
Merck Index | 15,8793 |
Solubility Information | Solubility in water: soluble |
Formula Weight | 216.02 |
Percent Purity | 98% |
Packaging | Glass bottle |
Chemical Name or Material | Sodium phosphite dibasic pentahydrate |
Safety and Handling
Product Identifier
- Sodium phosphite dibasic pentahydrate
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'