missing translation for 'onlineSavingsMsg'
Learn More

Sodium phosphite dibasic pentahydrate, 98%, Thermo Scientific Chemicals

Product Code. 11994281
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13517-23-2
Molecular Formula HNa2O35H2O
Molecular Weight (g/mol) 216.02
Synonym sodium phosphite dibasic pentahydrate, di-sodium hydrogen phosphite, di-sodium hydrogen phosphite pentahydrate, disodium pentahydrate hydrogen phosphite, 2na.ho3p.5h2o, di-sodiumhydrogenphosphitepentahydrate, disodium hydrogenphosphite pentahydrate, etidronate disodium related compound a, united states pharmacopeia usp reference standard

Specifications

Quantity 250 g
Assay Percent Range 98%
Merck Index 15,8793
Solubility Information Solubility in water: soluble
Formula Weight 216.02
Percent Purity 98%
Packaging Glass bottle
Chemical Name or Material Sodium phosphite dibasic pentahydrate
Product Identifier
  • Sodium phosphite dibasic pentahydrate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'