missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium phosphonoformate hexahydrate, 98+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
Description
Sodium phosphonoformate hexahydrate acts as an antiviral agent and reverse transcriptase inhibitor. It is also used to inhibit viral DNA polymerase. Further, it is used as a type II Pi transporter inhibitor.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Sodium phosphonoformate hexahydrate acts as an antiviral agent and reverse transcriptase inhibitor. It is also used to inhibit viral DNA polymerase. Further, it is used as a type II Pi transporter inhibitor.
Solubility
Soluble in water.
Notes
Incompatible with oxidizing agents.
Sodium phosphonoformate hexahydrate acts as an antiviral agent and reverse transcriptase inhibitor. It is also used to inhibit viral DNA polymerase. Further, it is used as a type II Pi transporter inhibitor.
Solubility
Soluble in water.
Notes
Incompatible with oxidizing agents.
Specifications
Quantity | 1 g |
Solubility Information | Soluble in water. |
IUPAC Name | trisodium;phosphonatoformate;hexahydrate |
Formula Weight | 300.08 (191.98 Anhydrous) |
Percent Purity | ≥98% |
Chemical Name or Material | Sodium phosphonoformate hexahydrate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'