Learn More
Sodium polyphosphate, pure, Thermo Scientific Chemicals
£24.52 - £145.73
Chemical Identifiers
CAS | 50813-16-6 |
---|---|
Molecular Formula | (NaO3P)n |
MDL Number | MFCD00134118 |
Synonym | hexasodium oxido oxido phosphonatooxy phosphoryl oxy phosphoryl oxy phospho phosphonato oxy phosphinate |
IUPAC Name | Sodium polyphosphate |
SMILES | [Na]OP(-*)(=O)O-* |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10055293
|
Thermo Scientific Acros
390932500 |
250 g | Plastic bottle |
£24.52
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10268893
|
Thermo Scientific Acros
390930010 |
1 kg | Plastic bottle |
£44.72
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10025533
|
Thermo Scientific Acros
390930050 |
5 kg | Plastic bottle |
£145.73
5kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
50813-16-6 | |
MFCD00134118 | |
Sodium polyphosphate |
(NaO3P)n | |
hexasodium oxido oxido phosphonatooxy phosphoryl oxy phosphoryl oxy phospho phosphonato oxy phosphinate | |
[Na]OP(-*)(=O)O-* |
Specifications
600.0°C | |
White | |
(NaO3P)n | |
MFCD00134118 | |
hexasodium oxido oxido phosphonatooxy phosphoryl oxy phosphoryl oxy phospho phosphonato oxy phosphinate | |
[Na]OP(-*)(=O)O-* | |
65 to 70% | |
Sodium polyphosphate |
50813-16-6 | |
Crystalline Powder | |
(NaPO3)n | |
15, 8773 | |
Solubility in water: 1000g/l (20°C) | |
Sodium polyphosphate | |
Pure |
Safety and Handling
GHS H Statement
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 256-779-4
- Sodium polyphosphate
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only