Learn More
Sodium pyrophosphate decahydrate, 99+%, for analysis, Thermo Scientific Chemicals
£22.31 - £356.78
Chemical Identifiers
CAS | 13472-36-1 |
---|---|
Molecular Formula | Na4O7P2·10H2O |
Molecular Weight (g/mol) | 446.05 |
MDL Number | MFCD00149200 |
InChI Key | VZWGHDYJGOMEKT-UHFFFAOYSA-J |
Synonym | sodium pyrophosphate decahydrate, unii-iy3dkb96qw, tetrasodium pyrophosphate decahydrate, diphosphoric acid, tetrasodium salt, decahydrate, iy3dkb96qw, pyrophosphate sodium salt decahydrate, pyrophosphate inorganic, colostrum, sodium phosphate pyro, sodium pyrophosphate na4p2o7 decahydrate |
PubChem CID | 3084150 |
IUPAC Name | tetrasodium;phosphonato phosphate;decahydrate |
SMILES | O.O.O.O.O.O.O.O.O.O.[O-]P(=O)([O-])OP(=O)([O-])[O-].[Na+].[Na+].[Na+].[Na+] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10002710
|
Thermo Scientific Acros
205970250 |
25 g |
£22.31
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10368580
|
Thermo Scientific Acros
205975000 |
500 g |
£34.47
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10144423
|
Thermo Scientific
205970025 |
2.5 kg |
£111.56
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15204684
|
Thermo Scientific Acros
205970100 |
10 kg |
£356.78
10kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Biological bufferChemical Identifiers
13472-36-1 | |
446.05 | |
VZWGHDYJGOMEKT-UHFFFAOYSA-J | |
3084150 | |
O.O.O.O.O.O.O.O.O.O.[O-]P(=O)([O-])OP(=O)([O-])[O-].[Na+].[Na+].[Na+].[Na+] |
Na4O7P2·10H2O | |
MFCD00149200 | |
sodium pyrophosphate decahydrate, unii-iy3dkb96qw, tetrasodium pyrophosphate decahydrate, diphosphoric acid, tetrasodium salt, decahydrate, iy3dkb96qw, pyrophosphate sodium salt decahydrate, pyrophosphate inorganic, colostrum, sodium phosphate pyro, sodium pyrophosphate na4p2o7 decahydrate | |
tetrasodium;phosphonato phosphate;decahydrate |
Specifications
93.8°C | |
13472-36-1 | |
Crystals | |
Na4O7P2·10H2O | |
MFCD00149200 | |
sodium pyrophosphate decahydrate, unii-iy3dkb96qw, tetrasodium pyrophosphate decahydrate, diphosphoric acid, tetrasodium salt, decahydrate, iy3dkb96qw, pyrophosphate sodium salt decahydrate, pyrophosphate inorganic, colostrum, sodium phosphate pyro, sodium pyrophosphate na4p2o7 decahydrate | |
VZWGHDYJGOMEKT-UHFFFAOYSA-J | |
tetrasodium;phosphonato phosphate;decahydrate | |
3084150 | |
≥99% | |
Glass Bottle | |
Sodium pyrophosphate decahydrate, p.a. |
80.0°C | |
White | |
99+% | |
Na4P2O7·10H2O | |
10,9063 | |
Solubility in water: 62g/L (20°C). Other solubilities: insoluble in ammonia,insoluble in alcohol | |
O.O.O.O.O.O.O.O.O.O.[O-]P(=O)([O-])OP(=O)([O-])[O-].[Na+].[Na+].[Na+].[Na+] | |
446.05 | |
446.05 | |
Analytical | |
1.82 |
Safety and Handling
GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
- Sodium pyrophosphate decahydrate
Signal Word
- Warning
Hazard Category
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
- MIXTURE LIST-Contains : Sodium pyrophosphatedecahydrate, Sodiumpyrophosphate
RUO – Research Use Only