missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Pyruvate (100 mM)

Sodium Pyruvate (100 mM)
Brand: Gibco 11360039
This item is not returnable.
View return policy
Description
Sodium Pyruvate is commonly added to cell culture medium as an energy source in addition to glucose. Since the cells make sodium pyruvate as an intermediate metabolite in the glycolysis pathway, it is not necessary to supplement all cultures. However, if the cells have been grown in medium supplemented with sodium pyruvate, we recommend the supplementation is continued as the cells become accustomed to this addition and their growth will lag without it. Gibco™ Sodium Pyruvate, 100 mM Solution is formulated as 11 g per liter of water. The final concentration of sodium pyruvate used in most cell culture medium is 1 mM.Product Use
For in vitro diagnostic use.
Dual-site cGMP Manufacturing and Quality System
Gibco™ Sodium Pyruvate Solution is manufactured at a cGMP compliant facility, located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard. For supply chain continuity, we offer an identical Gibco™ Sodium Pyruvate Solution product made in our Grand Island facility (11360-070). This facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standards.

Specifications
FDA, ISO 13485 | |
Sodium Pyruvate (100mM), 100mL, Liquid, 2 to 8°C Storage Temperature, 12 Months Shelf Life, Protect From Light, Used In Cell Culture and Mammalian Culture, Fewer Resources Used, Less Waste, For In-vitro Diagnostic Use | |
Sodium Pyruvate | |
100 mL |
2 to 8°C | |
Liquid | |
Mammalian Cell Culture | |
RUO - research use only |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 12
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- 3,4-Dichlorophenol
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
Hazard Statement
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
- MIXTURE LIST-Contains : 3,4-Dichlorophenol
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Sodium Pyruvate (100 mM) > 100mL
For In Vitro Diagnostic Use.
Spot an opportunity for improvement?Share a Content Correction