Learn More
Sodium selenate, anhydrous, 99.8+% (metals basis), Thermo Scientific Chemicals
For Use As a Reagent
£22.00 - £307.00
Chemical Identifiers
CAS | 13410-01-0 |
---|---|
Molecular Formula | Na2O4Se |
Molecular Weight (g/mol) | 188.95 |
MDL Number | MFCD00003490 |
InChI Key | MHQOTKLEMKRJIR-UHFFFAOYSA-L |
Synonym | sodium selenate, disodium selenate, natriumseleniat, selenic acid, disodium salt, caswell no. 791, natriumseleniat german, sel-tox sso2 and ss-20, unii-5dqp25600a, selenic acid h2seo4 , disodium salt, ccris 1259 |
PubChem CID | 25960 |
ChEBI | CHEBI:77775 |
IUPAC Name | disodium;selenate |
SMILES | [Na+].[Na+].[O-][Se]([O-])(=O)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11356459
|
Thermo Scientific Alfa Aesar
012613.06 |
5 g |
£22.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11326459
|
Thermo Scientific Alfa Aesar
012613.18 |
50 g |
£54.20
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11336459
|
Thermo Scientific Alfa Aesar
012613.30 |
250 g |
£181.00
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11346459
|
Thermo Scientific Alfa Aesar
012613.A1 |
1 kg |
£307.00
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Sodium selenate is used in glass industry, and produces red hue in glass. It acts as a decoloring agent in glass production. Combined with ferrous oxides, it produces colorless glass. Appropriate preparation of sodium selenate is used in multivitamins.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
13410-01-0 | |
188.95 | |
MHQOTKLEMKRJIR-UHFFFAOYSA-L | |
25960 | |
disodium;selenate |
Na2O4Se | |
MFCD00003490 | |
sodium selenate, disodium selenate, natriumseleniat, selenic acid, disodium salt, caswell no. 791, natriumseleniat german, sel-tox sso2 and ss-20, unii-5dqp25600a, selenic acid h2seo4 , disodium salt, ccris 1259 | |
CHEBI:77775 | |
[Na+].[Na+].[O-][Se]([O-])(=O)=O |
Specifications
32° (transition) | |
Purple | |
99.8+% (metals basis) | |
MFCD00003490 | |
Soluble in water. | |
[Na+].[Na+].[O-][Se]([O-])(=O)=O | |
188.95 | |
CHEBI:77775 | |
Odorless | |
White powder | |
Sodium selenate, anhydrous |
13410-01-0 | |
Powder | |
Na2O4Se | |
sodium selenate, disodium selenate, natriumseleniat, selenic acid, disodium salt, caswell no. 791, natriumseleniat german, sel-tox sso2 and ss-20, unii-5dqp25600a, selenic acid h2seo4 , disodium salt, ccris 1259 | |
MHQOTKLEMKRJIR-UHFFFAOYSA-L | |
disodium;selenate | |
25960 | |
188.94 | |
Hygroscopic | |
3.2 g/cm3 at 20°C |
Safety and Handling
P260-P264b-P270-P271-P301+P310-P304+P340-P311-P314-P330-P501c
H300-H331-H373
missing translation for 'dotInformation' : DOT Class: 6.1, Packaging Group: I
missing translation for 'einecsNumber' : 236-501-8
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Sodium selenate
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 1
- Acute toxicity Category 3
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
- H300-Fatal if swallowed.
- H331-Toxic if inhaled.
- H373-May cause damage to organs.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P311-Call a POISON CENTER/doctor/ .
- P330-Rinse mouth.
Supplemental information
- MIXTURE LIST-Contains : sodium selenate
RUO – Research Use Only