missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium selenite, 44-46% Se, anhydrous, Thermo Scientific Chemicals
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | 350°C |
Color | White to Beige |
Physical Form | Powder |
Quantity | 25 g |
Linear Formula | Na2SeO3 |
Merck Index | 15, 8802 |
Solubility Information | Solubility in water: 950g/L (20°C). Other solubilities: insoluble in alcohol |
Formula Weight | 172.94 |
Percent Purity | 44 to 46% Se |
Loss on Drying | 0.5% max. |
Show More |
Safety and Handling


Product Identifier
- Sodium selenite
- Danger
- Acute toxicity Category 2
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Skin sensitiser Category 1
- H300-Fatal if swallowed.
- H315-Causes skin irritation.
- H317-May cause an allergic skin reaction.
- H319-Causes serious eye irritation.
- H330-Fatal if inhaled.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- EUH031-Contact with acids liberates toxic gas.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Sodium selenite, 44-46% Se, anhydrous, Thermo Scientific Chemicals >
Spot an opportunity for improvement?Share a Content Correction