missing translation for 'onlineSavingsMsg'
Learn More

Sodium Silicate Solution, Reagent Grade, Honeywell™

£22.00 - £163.65

Chemical Identifiers

CAS 1344-09-8
Molecular Formula H10Na2O8Si
Molecular Weight (g/mol) 212.14
MDL Number MFCD00003492
InChI Key ADPGKKZKGXANON-UHFFFAOYSA-N
Synonym sodium metasilicate, sodium silicate, waterglass, water glass, sodium siliconate, sodium polysilicate, sodium sesquisilicate, sodium silicate glass, sodium silicate solution, sodium water glass
PubChem CID 23266
ChEBI CHEBI:60720
IUPAC Name disodium;dioxido(oxo)silane
SMILES O.O.O.O.O.[Na+].[Na+].[O-][Si]([O-])=O
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
15618370
View Documents
Honeywell Research Chemicals
338443-25ML
25 mL Poly bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15664090
View Documents
Honeywell Research Chemicals
338443-1L
1 L Poly bottle
£59.10
1 L
Please to purchase this item. Need a web account? Register with us today!
15674090
View Documents
Honeywell Research Chemicals
338443-3L
3 L Poly bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Specifications

Chemical Identifiers

1344-09-8
212.14
ADPGKKZKGXANON-UHFFFAOYSA-N
23266
disodium;dioxido(oxo)silane
H10Na2O8Si
MFCD00003492
sodium metasilicate, sodium silicate, waterglass, water glass, sodium siliconate, sodium polysilicate, sodium sesquisilicate, sodium silicate glass, sodium silicate solution, sodium water glass
CHEBI:60720
O.O.O.O.O.[Na+].[Na+].[O-][Si]([O-])=O

Specifications

1344-09-8
Na2O(SiO2)x · xH2O
UN3266
ADPGKKZKGXANON-UHFFFAOYSA-N
disodium;dioxido(oxo)silane
23266
1.39g/mL at 25°C
H10Na2O8Si
MFCD00003492
sodium metasilicate, sodium silicate, waterglass, water glass, sodium siliconate, sodium polysilicate, sodium sesquisilicate, sodium silicate glass, sodium silicate solution, sodium water glass
O.O.O.O.O.[Na+].[Na+].[O-][Si]([O-])=O
212.14
CHEBI:60720
Sodium silicate solution
Videos
Safety and Handling

Safety and Handling

P261-P280-P305 + P351 + P338-P310

missing translation for 'dotInformation' : Class 8/ PGIII

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.