missing translation for 'onlineSavingsMsg'
Learn More
Sodium Standard Metal Solution 10,000ppm, for ICP, Solution HNO3 1M, Fisher Chemical Product Code.: 10785521

Sodium Standard Metal Solution 10,000ppm, for ICP, Solution HNO3 1M, Fisher Chemical

Product Code. 10785521
100 mL, HDPE plastic bottle
Click to view available options
Quantity:
100 mL
Packaging:
HDPE plastic bottle
This item is not returnable. View return policy

Product Code. 10785521

Brand: Fisher Chemical J/8315/05

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Color Colorless
pH 1
MDL Number 85307
Quantity 100 mL
Packaging HDPE plastic bottle
Form Liquid
compliance-icons
Product Identifier
  • Sodium solution 10 000 ppm in 1M HNO6
Signal Word
  • Danger
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/A§.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sodium Standard Metal Solution 10,000ppm, for ICP, Solution HNO3 1M, Fisher Chemical >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.