missing translation for 'onlineSavingsMsg'
Learn More
Sodium sulfate, 99+%, ACS reagent, granular, anhydrous, Thermo Scientific Chemicals Product Code.: 10041501

Sodium sulfate, 99+%, ACS reagent, granular, anhydrous, Thermo Scientific Chemicals

Product Code. 10041501
1 kg, Plastic bottle
Click to view available options
Quantity:
1 kg
2.5 kg
10 kg
Packaging:
Plastic bottle
Plastic drum
This item is not returnable. View return policy

Product Code. 10041501

Brand: Thermo Scientific Acros 354250010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7757-82-6
Molecular Formula Na2O4S
Molecular Weight (g/mol) 142.04
MDL Number MFCD00003504
InChI Key PMZURENOXWZQFD-UHFFFAOYSA-L
Synonym sodium sulfate, disodium sulfate, salt cake, thenardite, sodium sulfate anhydrous, sodium sulphate, sodium sulfate, anhydrous, sulfuric acid disodium salt, disodium sulphate, sodium sulfate 2:1Show MoreShow Less
PubChem CID 24436
ChEBI CHEBI:32149
SMILES [Na+].[Na+].[O-]S([O-])(=O)=O

Specifications

Boiling Point >1700.0°C
Melting Point 884.0°C
Color Colorless to Orange
pH 5.2 to 7.0 (5% soln. at 25°C)
Physical Form Granules
Quantity 1 kg
Assay Percent Range 99+%
Linear Formula Na2SO4
Fieser 01,1103
Merck Index 15, 8809
IUPAC Name disodium sulfate
Formula Weight 142.04
Percent Purity ≥99%
Grade ACS Reagent
Identification Sodium : passes test (USP) Sulfate : passes test (USP)
Loss on Ignition 0.5 % max.
Packaging Plastic bottle
Decomposition Information Decomposition Information: >890°C
Density 2.68g/mL
Chemical Name or Material Sodium sulfate, Anhydrous
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0469275 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number354250025,354250010,354250100
Lot NumberA0464090 Certificate TypeCertificate of Analysis Date18/07/2024 Catalog Number354250025,354250010,354250100
Lot NumberB0155539 Certificate TypeCertificate of Analysis Date11/06/2024 Catalog Number354250025,354250010,354250100
Lot NumberB0148849 Certificate TypeCertificate of Analysis Date14/11/2023 Catalog Number354250025,354250010,354250100
Lot NumberB0150312 Certificate TypeCertificate of Analysis Date14/11/2023 Catalog Number354250025,354250010,354250100

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sodium sulfate, 99+%, ACS reagent, granular, anhydrous, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.