missing translation for 'onlineSavingsMsg'
Learn More

Sodium sulfate, ACS, 99.0% min

Product Code. 11397277
Click to view available options
Quantity:
500 g
2 kg
10 kg
Unit Size:
10kg
2kg
500g
This item is not returnable. View return policy

Product Code. 11397277

Brand: Thermo Scientific Alfa Aesar 011560.A3

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For Use in Drying organic Liquids, in Kjeldahl Nitrogen Determination, in Manufacturing of Glass | CAS: 7757-82-6 | Na2O4S | 142.04 g/mol

In drying organic liquids, in Kjeldahl nitrogen determination, in manufacturing of glass. Sodium sulfate anhydrous is used primarily for drying non-aqueous solvents by removing traces of water. It is used as a fining agent for molten glass, levelling agent in textile processing, and filler in detergents. It is also used in Kraft process for paper pulping, defrosting windows, manufacture of starch, glass, and soap.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7757-82-6
Molecular Formula Na2O4S
Molecular Weight (g/mol) 142.04
MDL Number MFCD00003504
InChI Key PMZURENOXWZQFD-UHFFFAOYSA-L
Synonym sodium sulfate, disodium sulfate, salt cake, thenardite, sodium sulfate anhydrous, sodium sulphate, sodium sulfate, anhydrous, sulfuric acid disodium salt, disodium sulphate, sodium sulfate 2:1
PubChem CID 24436
ChEBI CHEBI:32149
IUPAC Name disodium;sulfate
SMILES [Na+].[Na+].[O-]S([O-])(=O)=O

Specifications

Melting Point 884°C
Color White
Physical Form Powder
Quantity 2 kg
Assay Percent Range 99.0% min.
Solubility Information Soluble in water. Insoluble in alcoholSoluble in water,glycerol and hydrogen iodide. Insoluble in ethanol and many organic solvents.
Formula Weight 142.04
Grade ACS
Odor Odorless
Refractive Index 1.484
Sensitivity Hygroscopic
Appearance White powder
Density 2.68 g/cm3 at 20°C
Chemical Name or Material Sodium sulfate, For ACS analysis
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.