missing translation for 'onlineSavingsMsg'
Learn More
Sodium Sulfate, ACS Reagent, Anhydrous Powder, Honeywell Fluka™ Product Code.: 15642390

Sodium Sulfate, ACS Reagent, Anhydrous Powder, Honeywell Fluka™

Product Code. 15642390 Shop All Honeywell Fluka Products
2.5 kg
Click to view available options
Quantity:
25 g
500 g
1 kg
2.5 kg
5 kg
12 kg
This item is not returnable. View return policy

Product Code. 15642390

Brand: Honeywell-Fluka 2385972.5KG

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

ACS Reagent, anhydrous, powder,≥99.0%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7757-82-6
Molecular Formula Na2O4S
Molecular Weight (g/mol) 142.04
MDL Number MFCD00003504
InChI Key PMZURENOXWZQFD-UHFFFAOYSA-L
Synonym sodium sulfate, disodium sulfate, salt cake, thenardite, sodium sulfate anhydrous, sodium sulphate, sodium sulfate, anhydrous, sulfuric acid disodium salt, disodium sulphate, sodium sulfate 2:1Show MoreShow Less
PubChem CID 24436
ChEBI CHEBI:32149
IUPAC Name disodium;sulfate
SMILES [Na+].[Na+].[O-]S([O-])(=O)=O

Specifications

Physical Form Anhydrous Powder
Quantity 2.5 kg
Linear Formula Na2SO4
UN Number NONH for all modes of transport
Formula Weight 142.04g/mol
Percent Purity ≥99%
Grade ACS Reagent
Density 2.68g/mL at 25°C
Chemical Name or Material Sodium Sulfate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sodium Sulfate, ACS Reagent, Anhydrous Powder, Honeywell Fluka™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.