missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Sulfate Anhydrous, 99+%, Extra Pure, Fisher Chemical
44.31 GBP valid until 2024-08-31
Use promo code "21743" to get your promotional price.
Use promo code "21743" to get your promotional price.
Click to view available options
Quantity:
1 kg
Packaging:
Plastic powder jar
Specifications
Melting Point | 884°C |
CAS | 7757-82-6 |
Color | White |
pH | 5 to 8 |
Physical Form | Solid |
Quantity | 1 kg |
Molecular Formula | Na2O4S |
MDL Number | MFCD00003504 |
Synonym | sodium sulfate, disodium sulfate, salt cake, thenardite, sodium sulfate anhydrous, sodium sulphate, sodium sulfate, anhydrous, sulfuric acid disodium salt, disodium sulphate, sodium sulfate 2:1 |
InChI Key | PMZURENOXWZQFD-UHFFFAOYSA-L |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'