missing translation for 'onlineSavingsMsg'
Learn More
Sodium sulfate, anhydrous, 99%, Thermo Scientific Chemicals Product Code.: 11498717

Sodium sulfate, anhydrous, 99%, Thermo Scientific Chemicals

Product Code. 11498717
1000 g
Click to view available options
Quantity:
1000 g
5000 g
This item is not returnable. View return policy

Product Code. 11498717

Brand: Thermo Scientific Alfa Aesar A19890.0B

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sodium sulfate anhydrous is used primarily for drying non-aqueous solvents by removing traces of water. It is used as a fining agent for molten glass, levelling agent in textile processing, and filler in detergents. It is also used in Kraft process for paper pulping, defrosting windows, manufacture of starch, glass, and soap.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium sulfate anhydrous is used primarily for drying non-aqueous solvents by removing traces of water. It is used as a fining agent for molten glass, levelling agent in textile processing, and filler in detergents. It is also used in Kraft process for paper pulping, defrosting windows, manufacture of starch, glass, and soap.

Solubility
Soluble in water, glycerol and hydrogen iodide. Insoluble in ethanol and many organic solvents.

Notes
Anhydrous sodium sulfate is hygroscopic in nature, and absorbs moisture converting to the hydrated form. The hydrated form on heating gets converted back to anhydrous form.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7757-82-6
Molecular Formula Na2O4S
Molecular Weight (g/mol) 142.04
MDL Number MFCD00003504
InChI Key PMZURENOXWZQFD-UHFFFAOYSA-L
Synonym sodium sulfate, disodium sulfate, salt cake, thenardite, sodium sulfate anhydrous, sodium sulphate, sodium sulfate, anhydrous, sulfuric acid disodium salt, disodium sulphate, sodium sulfate 2:1
PubChem CID 24436
ChEBI CHEBI:32149
SMILES [Na+].[Na+].[O-]S([O-])(=O)=O

Specifications

Melting Point 884°C
Quantity 1000 g
Linear Formula Na2SO4
Merck Index 14,8680
Solubility Information Soluble in water,glycerol and hydrogen iodide. Insoluble in ethanol and many organic solvents.
IUPAC Name disodium;sulfate
Formula Weight 142.04
Percent Purity 99%
Odor Odorless
Refractive Index 1.484
Sensitivity Hygroscopic
Density 2.68
Chemical Name or Material Sodium sulfate, anhydrous
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sodium sulfate, anhydrous, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.