missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Sodium sulfate decahydrate, 99%, for biochemistry
£32.36 - £106.53
Chemical Identifiers
CAS | 7727-73-3 |
---|---|
Molecular Formula | Na2O4S·10H2O |
Molecular Weight (g/mol) | 322.2 |
InChI Key | RSIJVJUOQBWMIM-UHFFFAOYSA-L |
Synonym | sodium sulfate decahydrate, glauber's salt, disodium sulfate decahydrate, sodium sulfate usp, sulfuric acid disodium salt, decahydrate, unii-0ypr65r21j, sodium sulfate na2so4 decahydrate, sodium sulphate decahydrate, sodium sulfate decahydrate na2so4.10h2o, disodium sulfate decahydrate na2so4.10h2oShow More |
PubChem CID | 62649 |
ChEBI | CHEBI:32586 |
IUPAC Name | disodium;sulfate;decahydrate |
SMILES | O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[Na+].[Na+] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
12645127
|
Thermo Scientific Chemicals
447935000 |
500 g | Plastic bottle |
£32.36
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12672936
|
Thermo Scientific Chemicals
447930025 |
2.5 kg | Plastic bottle |
£106.53
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7727-73-3 | |
322.2 | |
sodium sulfate decahydrate, glauber's salt, disodium sulfate decahydrate, sodium sulfate usp, sulfuric acid disodium salt, decahydrate, unii-0ypr65r21j, sodium sulfate na2so4 decahydrate, sodium sulphate decahydrate, sodium sulfate decahydrate na2so4.10h2o, disodium sulfate decahydrate na2so4.10h2oShow More | |
CHEBI:32586 | |
O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[Na+].[Na+] |
Na2O4S·10H2O | |
RSIJVJUOQBWMIM-UHFFFAOYSA-L | |
62649 | |
disodium;sulfate;decahydrate |
Specifications
884.0°C | |
99% | |
Na2SO4·10H2O | |
Clear colorless | |
O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[Na+].[Na+] | |
322.2 | |
CHEBI:32586 | |
99% | |
Sodium sulfate decahydrate |
7727-73-3 | |
Na2O4S·10H2O | |
sodium sulfate decahydrate, glauber's salt, disodium sulfate decahydrate, sodium sulfate usp, sulfuric acid disodium salt, decahydrate, unii-0ypr65r21j, sodium sulfate na2so4 decahydrate, sodium sulphate decahydrate, sodium sulfate decahydrate na2so4.10h2o, disodium sulfate decahydrate na2so4.10h2o | |
RSIJVJUOQBWMIM-UHFFFAOYSA-L | |
disodium;sulfate;decahydrate | |
62649 | |
322.2 | |
Biochemical |
Safety and Handling
Product Identifier
- Sodium sulfate decahydrate
RUO – Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Thermo Scientific Chemicals Sodium sulfate decahydrate, 99%, for biochemistry