missing translation for 'onlineSavingsMsg'
Learn More
Sodium sulfate decahydrate, 99%, Thermo Scientific Chemicals Product Code.: 11457667

Sodium sulfate decahydrate, 99%, Thermo Scientific Chemicals

Product Code. 11457667
1000 g
Click to view available options
Quantity:
1000 g
25,000 g
5000 g
This item is not returnable. View return policy

Product Code. 11457667

Brand: Thermo Scientific Alfa Aesar A15702.0B

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sodium sulfate decahydrate is used as a laboratory reagent to precipitate protein. It is largely used as filler in powdered home laundry detergents. It facilitates in leveling, reducing negative charges on fibers, thereby allows dye to penetrate evenly. It avoids the corrosion of stainless steel vessels used in dyeing. It is also used as a laxative and effectively replaces the drug paracetamol in the body.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium sulfate decahydrate is used as a laboratory reagent to precipitate protein. It is largely used as filler in powdered home laundry detergents. It facilitates in leveling, reducing negative charges on fibers, thereby allows dye to penetrate evenly. It avoids the corrosion of stainless steel vessels used in dyeing. It is also used as a laxative and effectively replaces the drug paracetamol in the body.

Solubility
Soluble in water and glycerin. Insoluble in ethanol and methanol.

Notes
Stable. Incompatible with strong acids, aluminum and magnesium.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7727-73-3
Molecular Formula H20Na2O14S
Molecular Weight (g/mol) 322.186
MDL Number MFCD00149189
InChI Key RSIJVJUOQBWMIM-UHFFFAOYSA-L
Synonym sodium sulfate decahydrate, glauber's salt, disodium sulfate decahydrate, sodium sulfate usp, sulfuric acid disodium salt, decahydrate, unii-0ypr65r21j, sodium sulfate na2so4 decahydrate, sodium sulphate decahydrate, sodium sulfate decahydrate na2so4.10h2o, disodium sulfate decahydrate na2so4.10h2o
PubChem CID 62649
ChEBI CHEBI:32586
IUPAC Name disodium;sulfate;decahydrate
SMILES O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[Na+].[Na+]

Specifications

Melting Point ∼32°C
pH 5.2 to 8.0
Quantity 1000 g
Merck Index 14,8680
Solubility Information Soluble in water and glycerin. Insoluble in ethanol and methanol.
Formula Weight 322.19 (142.04 Anhydrous)
Percent Purity 99%
Odor Odorless
Density 1.46
Chemical Name or Material Sodium sulfate decahydrate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10249234 Certificate TypeCertificate of Analysis Date03/01/2024 Catalog NumberA15702.0B,A15702.0I,A15702.0G
Lot Number10241724 Certificate TypeCertificate of Analysis Date29/06/2022 Catalog NumberA15702.0B,A15702.0I,A15702.0G
Lot Number10239059 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog NumberA15702.0B,A15702.0I,A15702.0G
Lot Number10239992 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog NumberA15702.0B,A15702.0I,A15702.0G
Lot Number10110733 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA15702.0B,A15702.0I,A15702.0G

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sodium sulfate decahydrate, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.