missing translation for 'onlineSavingsMsg'
Learn More

Sodium sulfate decahydrate, extra pure, Thermo Scientific Chemicals

Product Code. 13498328
Click to view available options
Quantity:
250 g
1 kg
2.5 kg
25 kg
Packaging:
Fibre drum
Plastic bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7727-73-3
Molecular Formula Na2O410H2O
Molecular Weight (g/mol) 322.2
MDL Number MFCD00149189
InChI Key RSIJVJUOQBWMIM-UHFFFAOYSA-L
Synonym sodium sulfate decahydrate, glauber's salt, disodium sulfate decahydrate, sodium sulfate usp, sulfuric acid disodium salt, decahydrate, unii-0ypr65r21j, sodium sulfate na2so4 decahydrate, sodium sulphate decahydrate, sodium sulfate decahydrate na2so4.10h2o, disodium sulfate decahydrate na2so4.10h2o
PubChem CID 62649
ChEBI CHEBI:32586
IUPAC Name disodium;sulfate;decahydrate
SMILES O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[Na+].[Na+]

Specifications

Melting Point 884.0°C
Color White
pH 5.2 to 9.2 (5% soln.)
Physical Form Crystalline Powder and/or Crystals
Quantity 25 kg
Linear Formula Na2SO4·10H2O
Fieser 01,1103
Merck Index 14,868
Solubility Information Solubility in water: soluble
Formula Weight 322.2
Percent Purity ≥99.0%
Grade Extra Pure
Packaging Fibre drum
Chemical Name or Material Sodium sulfate decahydrate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'