missing translation for 'onlineSavingsMsg'
Learn More
Learn More

- Home
- Products
- Chemicals
- Salts and Inorganics
- Inorganic Salts
- Alkali Metal Salts
- Sodium sulfate, Honeywell
Favorite item to keep tabs on it!
Saved!
Go to My Lists
You have reached the limit of 350 favorited items.
Manage Your Favorites
Sodium sulfate, Honeywell
Product Code. 156668701 kg
Click to view available options
Quantity:
1 kg
5 kg
6 x 1 kg
This item is not returnable.
View return policy
Product Code. 15666870
Brand: Honeywell Research Chemicals 719621KG
This item is not returnable.
View return policy
Description
- Products with a guarantee certificate and/or suitable for the stated analytical application.

Specifications
Chemical Identifiers
CAS | 7757-82-6 |
Molecular Formula | Na2O4S |
Molecular Weight (g/mol) | 142.04 |
MDL Number | MFCD00003504 |
InChI Key | PMZURENOXWZQFD-UHFFFAOYSA-L |
Synonym | sodium sulfate, disodium sulfate, salt cake, thenardite, sodium sulfate anhydrous, sodium sulphate, sodium sulfate, anhydrous, sulfuric acid disodium salt, disodium sulphate, sodium sulfate 2:1Show MoreShow Less |
PubChem CID | 24436 |
ChEBI | CHEBI:32149 |
SMILES | [Na+].[Na+].[O-]S([O-])(=O)=O |
Specifications
Quantity | 1 kg |
Linear Formula | Na2SO4 |
UN Number | NONH for all modes of transport |
IUPAC Name | disodium;sulfate |
Formula Weight | 142.04g/mol |
Percent Purity | ≥99% (T) |
Density | 2.68g/mL at 25°C |
Chemical Name or Material | Sodium sulfate |
Product Suggestions
You may be interested in these alternatives
-
Product Code. 10746292
-
-
-
-
Product Code. 10032580
Customers who viewed this item also viewed This information does not imply a recommendation or representation of any kind and any action taken upon the information provided is strictly at your own risk. The products shown have not been qualified by Fisher Scientific for any particular purpose, application or use and it is the responsibility of the purchaser to ensure that the performance of the product is suitable for the purchasers specific use or intended application.
Viewing 1-5 of 6
-
Catalog No. 10703811
£258.00 / 500g -
Catalog No. 10793611
£101.00 / 1kg -
Catalog No. 10117113
£25.71 / 100g -
Catalog No. 10112640
£50.40 / 1kg -
Catalog No. 10092740
£21.51 / 1kg -
Catalog No. 10401841
£22.81 / 500mL
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Sodium sulfate, Honeywell >
Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.
Spot an opportunity for improvement?Share a Content Correction