missing translation for 'onlineSavingsMsg'
Learn More

Sodium sulfate, Honeywell

Product Code. 15686870
6 x 1 kg
Click to view available options
Quantity:
1 kg
5 kg
6 x 1 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

Purum p.a., anhydrous,≥99.0% (T)

  • Products with a guarantee certificate and/or suitable for the stated analytical application.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7757-82-6
Molecular Formula Na2O4S
Molecular Weight (g/mol) 142.04
MDL Number MFCD00003504
InChI Key PMZURENOXWZQFD-UHFFFAOYSA-L
Synonym sodium sulfate, disodium sulfate, salt cake, thenardite, sodium sulfate anhydrous, sodium sulphate, sodium sulfate, anhydrous, sulfuric acid disodium salt, disodium sulphate, sodium sulfate 2:1
PubChem CID 24436
ChEBI CHEBI:32149
SMILES [Na+].[Na+].[O-]S([O-])(=O)=O

Specifications

Quantity 6 x 1 kg
Linear Formula Na2SO4
UN Number NONH for all modes of transport
IUPAC Name disodium sulfate
Formula Weight 142.04
Percent Purity ≥99.0% (T)
Density 2.68g/mL at 25°C
Chemical Name or Material Sodium sulfate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sodium sulfate, Honeywell >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.