missing translation for 'onlineSavingsMsg'
Learn More

Sodium Sulfate, MP Biomedicals™

Product Code. 11464662 Shop All MP Biomedicals Products
500 g
Click to view available options
Quantity:
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Used in protein crystallization and to investigate prion protein folding. Sodium Sulfate, MP Biomedicals is the sodium salt of sulfuric acid. It is a reagent used in large-scale applications such as the dyeing and printing of textiles and in the manufactu

TRUSTED_SUSTAINABILITY

Specifications

CAS 7757-82-6
Physical Form Powder
Quantity 500 g
Molecular Formula Na2O4S
MDL Number MFCD00003504
Synonym sodium sulfate, disodium sulfate, salt cake, thenardite, sodium sulfate anhydrous, sodium sulphate, sodium sulfate, anhydrous, sulfuric acid disodium salt, disodium sulphate, sodium sulfate 2:1
Solubility Information Soluble in water (100mg/mL), yielding a clear, colorless solution.
InChI Key PMZURENOXWZQFD-UHFFFAOYSA-L
SMILES [Na+].[Na+].[O-]S([O-])(=O)=O
IUPAC Name disodium;sulfate
Molecular Weight (g/mol) 142.04
PubChem CID 24436
ChEBI CHEBI:32149
Formula Weight 142.04
Percent Purity >99.0%
Grade ACS
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sodium Sulfate, MP Biomedicals™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.