missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium sulfide, 90+%, pure, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
50 g
250 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Color | Beige-Yellow |
Physical Form | Lump/Powder |
Quantity | 10 g |
Assay Percent Range | 90% min. (Iodimetry) |
Linear Formula | Na2S |
Merck Index | 15,881 |
Solubility Information | Solubility in water: 186g/L (20°C). Other solubilities: slightly soluble in alcohol, insoluble in ether |
IUPAC Name | disodium;sulfanide |
Formula Weight | 78.04 |
Percent Purity | ≥90% |
Show More |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-5 of 8
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0462007 | Certificate of Analysis | 19/04/2024 | 310180100,310182500,310180500 |
A0455017 | Certificate of Analysis | 21/09/2023 | 310180100,310182500,310180500 |
A0432728 | Certificate of Analysis | 25/08/2023 | 310180100,310182500,310180500 |
A0439703 | Certificate of Analysis | 02/05/2023 | 310180100,310182500,310180500 |
A0410221 | Certificate of Analysis | 13/12/2022 | 310180100,310182500,310180500 |
Safety and Handling



Product Identifier
- Sodium sulfide
- Danger
- Substances and mixtures corrosive to metals Category 1
- Acute toxicity Category 3
- Skin corrosion/irritation Category 1 B
- Serious eye damage/eye irritation Category 1
- ACUTE AQUATIC Acute 1
- H290-May be corrosive to metals.
- H301-Toxic if swallowed.
- H311-Toxic in contact with skin.
- H314-Causes severe skin burns and eye damage.
- H400-Very toxic to aquatic life.
- EUH031-Contact with acids liberates toxic gas.
- EUH071-Corrosive to the respiratory tract.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Sodium sulfide, 90+%, pure, Thermo Scientific Chemicals >
Spot an opportunity for improvement?Share a Content Correction