missing translation for 'onlineSavingsMsg'
Learn More

Sodium sulfide hydrate, 60-64%, extra pure, flakes, Thermo Scientific Chemicals

Product Code. 10294990
Click to view available options
Quantity:
1 kg
2.5 kg
10 kg
Packaging:
Fibre drum
Plastic bottle
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 27610-45-3
Molecular Formula Na2S
Molecular Weight (g/mol) 78.04
MDL Number MFCD00149183
InChI Key GRVFOGOEDUUMBP-UHFFFAOYSA-N
PubChem CID 45051681
SMILES [Na+].[Na+].[S--]

Specifications

Color Yellow
Physical Form Flakes and Chunks
Quantity 2.5 kg
Assay Percent Range 60 to 64% (Na2S) (Iodimetry)
Linear Formula Na2S·xH2O
Solubility Information Solubility in water: soluble. Other solubilities: slightly soluble in alcohol,insoluble in ether
IUPAC Name disodium;sulfanide;hydrate
Formula Weight 78.04
Percent Purity 60 to 64%
Grade Extra Pure
Packaging Plastic bottle
Chemical Name or Material Sodium sulfide hydrate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'