missing translation for 'onlineSavingsMsg'
Learn More

Sodium sulfite, 98.5%, for analysis, anhydrous

Product Code. 10117743 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
250 g
1 kg
5 kg
Packaging:
Glass Bottle
Plastic bottle
Unit Size:
1kg
250g
25g
5kg
This item is not returnable. View return policy

Product Code. 10117743

Brand: Thermo Scientific Acros 219270050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7757-83-7 | Na2O3S | 126.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7757-83-7
Molecular Formula Na2O3S
Molecular Weight (g/mol) 126.04
MDL Number MFCD00003503
InChI Key GEHJYWRUCIMESM-UHFFFAOYSA-L
Synonym sodium sulfite, disodium sulfite, sodium sulphite, sodium sulfite anhydrous, sulfurous acid, disodium salt, sulftech, natrium sulfurosum, sodiumsulfite, natriumsulfid, natriumsulfit
PubChem CID 24437
ChEBI CHEBI:86477
IUPAC Name disodium;sulfite
SMILES [O-]S(=O)[O-].[Na+].[Na+]

Specifications

Melting Point >500.0°C
Color White
Physical Form Fine Crystals or Crystalline Powder
Quantity 5 kg
Assay Percent Range 98% min. (Iodimetry)
Linear Formula Na2SO3
Fieser 02,387
Merck Index 15, 8811
Solubility Information Solubility in water: 230g/mL (20°C). Other solubilities: soluble in glycerol,ppractically insoluble in alcohol
Formula Weight 126.04
Percent Purity 98.5%
Grade Analytical
Packaging Plastic bottle
Specific Gravity 2.63
Density 2.6300g/mL
Chemical Name or Material Sodium sulfite, Anhydrous, For analysis
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.