missing translation for 'onlineSavingsMsg'
Learn More
Sodium sulfite, 98+%, ACS reagent, anhydrous, Thermo Scientific Chemicals Product Code.: 10464135

Sodium sulfite, 98+%, ACS reagent, anhydrous, Thermo Scientific Chemicals

Product Code. 10464135 Shop All Thermo Scientific Products
250 g, Plastic bottle
Click to view available options
Quantity:
250 g
1 kg
2.5 kg
Packaging:
Plastic bottle
This item is not returnable. View return policy

Product Code. 10464135

Brand: Thermo Scientific Acros 424432500

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7757-83-7
Molecular Formula Na2O3S
Molecular Weight (g/mol) 126.04
MDL Number MFCD00003503
InChI Key GEHJYWRUCIMESM-UHFFFAOYSA-L
Synonym sodium sulfite, disodium sulfite, sodium sulphite, sodium sulfite anhydrous, sulfurous acid, disodium salt, sulftech, natrium sulfurosum, sodiumsulfite, natriumsulfid, natriumsulfitShow MoreShow Less
PubChem CID 24437
ChEBI CHEBI:86477
IUPAC Name disodium;sulfite
SMILES [O-]S(=O)[O-].[Na+].[Na+]

Specifications

Melting Point >500.0°C
Color White
Physical Form Fine Crystalline Powder
Quantity 250 g
Assay Percent Range 98% min. (Iodimetry)
Linear Formula Na2SO3
Fieser 02,387
Merck Index 15, 8811
Solubility Information Solubility in water: 230 g/mL (20°C). Other solubilities: soluble in glycerol, ppractically insoluble in alcohol
Formula Weight 126.04
Percent Purity ≥98%
Grade ACS Reagent
Packaging Plastic bottle
Specific Gravity 2.63
Decomposition Information >500°C
Density 2.6300g/mL
Chemical Name or Material Sodium sulfite
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0467320 Certificate TypeCertificate of Analysis Date08/11/2024 Catalog Number424430010,424430025,424432500
Lot NumberA0445373 Certificate TypeCertificate of Analysis Date31/08/2023 Catalog Number424430010,424430025,424432500
Lot NumberA0437493 Certificate TypeCertificate of Analysis Date12/01/2023 Catalog Number424430010,424430025,424432500
Lot NumberA0399289 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number424430010,424430025,424432500
Lot NumberA0405740 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number424430010,424430025,424432500

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sodium sulfite, 98+%, ACS reagent, anhydrous, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.