missing translation for 'onlineSavingsMsg'
Learn More
Sodium sulfite, ACS, 98.0% min, Thermo Scientific Chemicals Product Code.: 11316818

Sodium sulfite, ACS, 98.0% min, Thermo Scientific Chemicals

Product Code. 11316818
10 kg
Click to view available options
Quantity:
500 g
10 kg
This item is not returnable. View return policy

Product Code. 11316818

Brand: Thermo Scientific Alfa Aesar 013454.A9

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sodium sulfite is used in the photography industry to protect developing solutions from oxidation; in the pulp and paper industry; in water treatment as an oxygen scavenger agent, as a desulfurizing and dechlorinating agent in the leather industry and as a bleaching agent in the textile industry.

Sodium sulfite is used in the photography industry to protect developing solutions from oxidation; in the pulp and paper industry; in water treatment as an oxygen scavenger agent, as a desulfurizing and dechlorinating agent in the leather industry and as a bleaching agent in the textile industry. It is used as a preservative to prevent dried fruit from discoloring. It finds application as a sulfonation and sulfomethylation agent in the chemical industry. It is involved in the production of sodium thiosulfate. Other applications include forth flotation ores, oil recovery and manufacture of dyes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7757-83-7
Molecular Formula Na2O3S
Molecular Weight (g/mol) 126.037
MDL Number MFCD00003503
InChI Key GEHJYWRUCIMESM-UHFFFAOYSA-L
Synonym sodium sulfite, disodium sulfite, sodium sulphite, sodium sulfite anhydrous, sulfurous acid, disodium salt, sulftech, natrium sulfurosum, sodiumsulfite, natriumsulfid, natriumsulfitShow MoreShow Less
PubChem CID 24437
ChEBI CHEBI:86477
IUPAC Name disodium;sulfite
SMILES [O-]S(=O)[O-].[Na+].[Na+]

Specifications

Melting Point >500°C (decomposition)
Physical Form Granules
Quantity 10 kg
Solubility Information Soluble in water and glycerol. Insoluble in alcohol,liquid chlorine and liquid ammonia.
Formula Weight 126.04
Percent Purity ≥98%
Grade ACS
Packaging Plastic bottle
Sensitivity Air and moisture sensitive
Specific Gravity 2.633
Density 2.633
Chemical Name or Material Sodium sulfite
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberZ19K024 Certificate TypeCertificate of Analysis Date21/01/2025 Catalog Number013454.A3,013454.36,013454.A9
Lot NumberW24K030 Certificate TypeCertificate of Analysis Date03/10/2024 Catalog Number013454.A3,013454.36,013454.A9
Lot NumberU27K033 Certificate TypeCertificate of Analysis Date24/09/2024 Catalog Number013454.A3,013454.36,013454.A9
Lot NumberP27K007 Certificate TypeCertificate of Analysis Date04/04/2024 Catalog Number013454.A3,013454.36,013454.A9
Lot NumberW06J053 Certificate TypeCertificate of Analysis Date21/09/2023 Catalog Number013454.A3,013454.36,013454.A9

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sodium sulfite, ACS, 98.0% min, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.