missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Sulfite Anhydrous, Extra Pure, SLR, Fisher Chemical™
Click to view available options
Quantity:
500 g
1 kg
Description
Sodium Sulfite Anhydrous, Extra Pure, SLR
Specifications
Specifications
Melting Point | 500°C |
Color | White |
pH | 8.5 to 10 |
Physical Form | Solid |
Quantity | 500 g |
Formula Weight | 126.04g/mol |
Packaging | Plastic powder jar |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'