missing translation for 'onlineSavingsMsg'
Learn More

Sodium Sulfite Anhydrous, Extra Pure, SLR, Fisher Chemical™

Product Code. 10386050
Click to view available options
Quantity:
500 g
1 kg

Sodium Sulfite Anhydrous, Extra Pure, SLR

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7757-83-7
Molecular Formula Na2O3S
Molecular Weight (g/mol) 126.037
MDL Number 3503
InChI Key GEHJYWRUCIMESM-UHFFFAOYSA-L
Synonym sodium sulfite, disodium sulfite, sodium sulphite, sodium sulfite anhydrous, sulfurous acid, disodium salt, sulftech, natrium sulfurosum, sodiumsulfite, natriumsulfid, natriumsulfit
PubChem CID 24437
ChEBI CHEBI:86477
IUPAC Name disodium;sulfite
SMILES [O-]S(=O)[O-].[Na+].[Na+]

Specifications

Melting Point 500°C
Color White
pH 8.5 to 10
Physical Form Solid
Quantity 1 kg
Formula Weight 126.04g/mol
Packaging Plastic powder jar
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'