missing translation for 'onlineSavingsMsg'
Learn More
Learn More

- Home
- Products
- Chemicals
- Salts and Inorganics
- Inorganic Salts
- Alkali Metal Salts
- Sodium sulphate, BAKER ANALYZED™, J.T. Baker™
Favorite item to keep tabs on it!
Saved!
Go to My Lists
You have reached the limit of 350 favorited items.
Manage Your Favorites
Sodium sulphate, BAKER ANALYZED™, J.T. Baker™
Product Code. 105788301 kg
Click to view available options
Quantity:
500 g
1 kg
5 kg
This item is not returnable.
View return policy
Product Code. 10578830
Brand: JT Baker 0313.1000
69.82 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Explore more special offers
Use promo code "25339" to get your promotional price.
Explore more special offers
This item is not returnable.
View return policy
Specifications
Chemical Identifiers
CAS | 7757-82-6 |
Molecular Formula | Na2O4S |
Molecular Weight (g/mol) | 142.04 |
MDL Number | MFCD00003504 |
InChI Key | PMZURENOXWZQFD-UHFFFAOYSA-L |
Synonym | Sulphuric acid disodium salt , Bisodium sulphate , Disodium sulphate |
IUPAC Name | disodium sulfate |
SMILES | [Na+].[Na+].[O-]S([O-])(=O)=O |
Specifications
Boiling Point | 1429°C (1013 hPa) |
Melting Point | 888°C |
pH | 5.2 to 9.2 (5% Solution at 25°C) |
Quantity | 1 kg |
Assay Percent Range | min. 99.0% |
Loss on Ignition | max. 0.5% |
Packaging | Plastic bottle for solids |
Density | 2.7 g/cm³ (20°C) |
Product Suggestions
You may be interested in these alternatives
-
Product Code. 10746292
-
-
-
-
Customers who viewed this item also viewed This information does not imply a recommendation or representation of any kind and any action taken upon the information provided is strictly at your own risk. The products shown have not been qualified by Fisher Scientific for any particular purpose, application or use and it is the responsibility of the purchaser to ensure that the performance of the product is suitable for the purchasers specific use or intended application.
Viewing 1-5 of 6
-
Catalog No. 10703811
£258.00 / 500g -
Catalog No. 10793611
£101.00 / 1kg -
Catalog No. 10117113
£25.71 / 100g -
Catalog No. 10112640
£50.40 / 1kg -
Catalog No. 10092740
£21.51 / 1kg -
Catalog No. 10401841
£22.81 / 500mL
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Sodium sulphate, BAKER ANALYZED™, J.T. Baker™ >
Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.
Spot an opportunity for improvement?Share a Content Correction